Anti-CUEDC2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverandør: Bioss
BS-4608R-A647
BOSSBS-4608R-A647EA 10450 NOK
BOSSBS-4608R-A647
Anti-CUEDC2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
Down-regulates ESR1 protein levels through the ubiquitination-proteasome pathway, regardless of the presence of 17 beta-estradiol. Also involved in 17 beta-estradiol-induced ESR1 degradation. Controls PGR protein levels through a similar mechanism.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CUEDC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol CUEDC2
Environmentally Preferable
Antigen-navn CUE Domain Containing 2
Antigen synonymer CUE domain-containing protein 2|C10orf66|bA18I14.5|CUEDC2|HOYS6
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 647
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 79004
Isotype IgG
ImmunoKjemi Yes
Immunogen 230-280/287
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR