Anti-AFP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-4708R-A680
BOSSBS-4708R-A680EA 10425 NOK
BOSSBS-4708R-A680
Anti-AFP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Alpha 1 Fetoprotein is a major plasma protein produced by the yolk sac and the liver during foetal life. Alpha fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistance of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the foetal counterpart of serum albumin, and the alpha fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. Alpha fetoprotein is found in monomeric as well as dimeric and trimeric forms, and binds copper,More Product Information