Anti-BMP15 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverandør: Bioss
BS-4720R-A488
BOSSBS-4720R-A488EA 10450 NOK
BOSSBS-4720R-A488
Anti-BMP15 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Required for ovarian folliculogenesis. Promotes primordial follicle development. Stimulates granulosa cell proliferation. Promotes cell transition from G0/G1 to S and G2/M phases, through an increase of CCND1 and CCNE1 expression, and RB1 phosphorylation. It regulates STAR expression and cAMP-dependent progesterone release in granulosa and thecal cells. Attenuates the suppressive effects of activin A on STAR expression and progesterone production by increasing the expression of inhibin B. It suppresses FST and FSTL3 production in granulosa-lutein cells.
Recommended Dilutions: IF(IHC-P): 1:More Product Information