Anti-GDF9 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-4720R-A750
BOSSBS-4720R-A750EA 10425 NOK
BOSSBS-4720R-A750
Anti-GDF9 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Required for ovarian folliculogenesis. Promotes primordial follicle development. Stimulates granulosa cell proliferation. Promotes cell transition from G0/G1 to S and G2/M phases, through an increase of CCND1 and CCNE1 expression, and RB1 phosphorylation. It regulates STAR expression and cAMP-dependent progesterone release in granulosa and thecal cells. Attenuates the suppressive effects of activin A on STAR expression and progesterone production by increasing the expression of inhibin B. It suppresses FST and FSTL3 production in granulosa-lutein cells.

Type: Primary
Antigen: GDF9
Clonality:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR