Anti-BMP15 Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-4720R-CY3
BOSSBS-4720R-CY3EA 10550 NOK
BOSSBS-4720R-CY3
Anti-BMP15 Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
Required for ovarian folliculogenesis. Promotes primordial follicle development. Stimulates granulosa cell proliferation. Promotes cell transition from G0/G1 to S and G2/M phases, through an increase of CCND1 and CCNE1 expression, and RB1 phosphorylation. It regulates STAR expression and cAMP-dependent progesterone release in granulosa and thecal cells. Attenuates the suppressive effects of activin A on STAR expression and progesterone production by increasing the expression of inhibin B. It suppresses FST and FSTL3 production in granulosa-lutein cells.

Recommended Dilutions: IF(IHC-P): 1:More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol BMP15
Environmentally Preferable
Antigen-navn Bone Morphogenetic Protein 15
Antigen synonymer ODG2|BMP-15|GDF9B|POF4|Bone morphogenetic protein 15
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy3®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 2661
Isotype IgG
ImmunoKjemi Yes
Immunogen 321-371/454
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR