Anti-TREM1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-4886R-A680
BOSSBS-4886R-A680EA 10425 NOK
BOSSBS-4886R-A680
Anti-TREM1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Stimulates neutrophil and monocyte-mediated inflammatory responses. Triggers release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. Amplifier of inflammatory responses that are triggered by bacterial and fungal infections and is a crucial mediator of septic shock (By similarity).

Type: Primary
Antigen: TREM1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 65-150/230
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR