Anti-TREM1 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-4886R-CY5
BOSSBS-4886R-CY5EA 10550 NOK
BOSSBS-4886R-CY5
Anti-TREM1 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Stimulates neutrophil and monocyte-mediated inflammatory responses. Triggers release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. Amplifier of inflammatory responses that are triggered by bacterial and fungal infections and is a crucial mediator of septic shock (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TREM1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol TREM1
Environmentally Preferable
Antigen-navn Triggering Receptor Expressed on Myeloid Cells 1
Antigen synonymer Trem1|CD354|TREM-1|Triggering receptor expressed on myeloid cells 1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5®
Reaktivitet Rat,Mouse
Vert Rabbit
Gen-ID 58217
Isotype IgG
ImmunoKjemi Yes
Immunogen 80-130/230
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR