Anti-NMRK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-5058R-A555
BOSSBS-5058R-A555EA 10450 NOK
BOSSBS-5058R-A555
Anti-NMRK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
ECHS1 (Enoyl Coenzyme A hydratase, short chain, 1) catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs, in the second step of the mitochondrial fatty acid beta-oxidation pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMRK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol NMRK1
Environmentally Preferable
Antigen-navn Nicotinamide Riboside Kinase 1
Antigen synonymer NRK1|bA235O14.2|C9orf95
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 555
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 54981
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR