Anti-ECHS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-5058R-A750
BOSSBS-5058R-A750EA 10425 NOK
BOSSBS-5058R-A750
Anti-ECHS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
ECHS1 (Enoyl Coenzyme A hydratase, short chain, 1) catalyses the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs, in the second step of the mitochondrial fatty acid beta-oxidation pathway.

Type: Primary
Antigen: ECHS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 111-210/290
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR