Anti-PAK6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverandør: Bioss
BS-5556R-A350
BOSSBS-5556R-A350EA 10800 NOK
BOSSBS-5556R-A350
Anti-PAK6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
This gene encodes a protein that shares a high degree of sequence similarity with p21-activated kinase (PAK) family members. The proteins of this family are Rac/Cdc42-associated Ste20-like Ser/Thr protein kinases, characterized by a highly conserved amino-terminal Cdc42/Rac interactive binding (CRIB) domain and a carboxyl-terminal kinase domain. PAK kinases are implicated in the regulation of a number of cellular processes, including cytoskeleton rearrangement, apoptosis and the MAP kinase signaling pathway. The protein encoded by this gene was found to interact with androgen receptor (AR),More Product Information
Bestill nå
SPESIFIKASJONER
Antigensymbol PAK6
Environmentally Preferable
Antigen-navn p21 Protein (Cdc42/Rac)-activated kinase 6
Antigen synonymer p21-activated protein kinase 6|p21-activated kinase 6|Serine/threonine-protein kinase PAK 6
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 350
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 56924
Isotype IgG
ImmunoKjemi Yes
Modifikasjon phospho-Ser165
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C