Anti-PAK6 Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-5556R-CY7
BOSSBS-5556R-CY7EA 10900 NOK
BOSSBS-5556R-CY7
Anti-PAK6 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
This gene encodes a protein that shares a high degree of sequence similarity with p21-activated kinase (PAK) family members. The proteins of this family are Rac/Cdc42-associated Ste20-like Ser/Thr protein kinases, characterized by a highly conserved amino-terminal Cdc42/Rac interactive binding (CRIB) domain and a carboxyl-terminal kinase domain. PAK kinases are implicated in the regulation of a number of cellular processes, including cytoskeleton rearrangement, apoptosis and the MAP kinase signaling pathway. The protein encoded by this gene was found to interact with androgen receptor (AR),More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol PAK6
Environmentally Preferable
Antigen-navn p21 Protein (Cdc42/Rac)-activated kinase 6
Antigen synonymer p21-activated protein kinase 6|p21-activated kinase 6|Serine/threonine-protein kinase PAK 6
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy7®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 56924
Isotype IgG
ImmunoKjemi Yes
Modifikasjon phospho-Ser165
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR