Anti-RGS19 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-5686R
BOSSBS-5686REA 8700 NOK
BOSSBS-5686R
Anti-RGS19 Rabbit Polyclonal Antibody
Antistoffer
G proteins mediate a number of cellular processes. The protein encoded by this gene belongs to the RGS (regulators of G protein signaling) family and specifically interacts with G protein, GAI3. This protein is a guanosine triphosphatase activating protein that functions to down regulate Galpha i/Galpha q linked signaling.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: RGS19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR