Anti-RGS19 Ser24 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-5687R-A680
BOSSBS-5687R-A680EA 10800 NOK
BOSSBS-5687R-A680
Anti-RGS19 Ser24 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
G proteins mediate a number of cellular processes. The protein encoded by this gene belongs to the RGS (regulators of G protein signaling) family and specifically interacts with G protein, GAI3. This protein is a guanosine triphosphatase activating protein that functions to down regulate Galpha i/Galpha q linked signaling.

Type: Primary
Antigen: RGS19 Ser24
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR