Anti-RGS19 Rabbit Polyclonal Antibody (Cy5.5®)

Leverandør: Bioss
BS-5687R-CY5.5
BOSSBS-5687R-CY5.5EA 10900 NOK
BOSSBS-5687R-CY5.5
Anti-RGS19 Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
G proteins mediate a number of cellular processes. The protein encoded by this gene belongs to the RGS (regulators of G protein signaling) family and specifically interacts with G protein, GAI3. This protein is a guanosine triphosphatase activating protein that functions to down regulate Galpha i/Galpha q linked signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RGS19
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol RGS19
Antigen-navn Regulator of G-protein Signaling 19
Environmentally Preferable
Antigen synonymer GAIP protein|GAIP|GNAI3IP|RGS19|Regulator of G-protein signaling 19|GS19|G-alpha interacting protein
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5.5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 10287
Isotype IgG
ImmunoKjemi Yes
Modifikasjon phospho-Ser24
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR