Anti-RGS19 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-5689R-A488
BOSSBS-5689R-A488EA 15800 NOK
BOSSBS-5689R-A488
Anti-RGS19 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
G proteins mediate a number of cellular processes. The protein encoded by this gene belongs to the RGS (regulators of G protein signaling) family and specifically interacts with G protein, GAI3. This protein is a guanosine triphosphatase activating protein that functions to down regulate Galpha i/Galpha q linked signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RGS19
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR