Anti-ARHGAP24 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Leverandør: Bioss
BS-5785R-A555
BOSSBS-5785R-A555EA 10450 NOK
BOSSBS-5785R-A555
Anti-ARHGAP24 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Rho GTPase-activating protein involved in cell polarity, cell morphology and cytoskeletal organization. Acts as a GTPase activator for the Rac-type GTPase by converting it to an inactive GDP-bound state. Controls actin remodeling by inactivating Rac downstream of Rho leading to suppress leading edge protrusion and promotes cell retraction to achieve cellular polarity. Able to suppress RAC1 and CDC42 activity in vitro. Overexpression induces cell rounding with partial or complete disruption of actin stress fibers and formation of membrane ruffles, lamellipodia, and filopodia. Isoform 2 is aMore Product Information