Anti-ELMO1 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-5807R
BOSSBS-5807REA 8500 NOK
BOSSBS-5807R
Anti-ELMO1 Rabbit Polyclonal Antibody
Antistoffer
ELMO1 is involved in cytoskeletal rearrangements required for phagocytosis of apoptotic cells and cell motility. ELMO1 acts in association with DOCK1 and CRK and was initially proposed to be required in complex with DOCK1 to activate Rac Rho small GTPases. It is belived to enhance the guanine nucleotide exchange factor (GEF) activity of DOCK1.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ELMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol ELMO1
Environmentally Preferable
Antigen-navn engulfment and cell motility 1
Antigen synonymer MGC126406|ced-12 homolog 1|ELMO-1|engulfment and cell motility 1|KIAA0281|CED12|CED-12
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 9844
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A.
Konsentrasjon 1 ug/ul
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR