Anti-PDCL Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-5812R-A350
BOSSBS-5812R-A350EA 10450 NOK
BOSSBS-5812R-A350
Anti-PDCL Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
Isoform 1: Functions as a co-chaperone for CCT in the assembly of heterotrimeric G protein complexes, facilitates the assembly of both Gbeta-Ggamma and RGS-Gbeta5 heterodimers. Isoform 2: Acts as a negative regulator of heterotrimeric G proteins assembly by trapping the preloaded G beta subunits inside the CCT chaperonin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDCL
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol PDCL
Antigen-navn Phosducin-Like
Environmentally Preferable
Antigen synonymer PHLOP1|PhLP
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 350
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 5082
Isotype IgG
ImmunoKjemi Yes
Immunogen 60-110/301
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR