Anti-PHLP Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-5812R-A750
BOSSBS-5812R-A750EA 10425 NOK
BOSSBS-5812R-A750
Anti-PHLP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Isoform 1: Functions as a co-chaperone for CCT in the assembly of heterotrimeric G protein complexes, facilitates the assembly of both Gbeta-Ggamma and RGS-Gbeta5 heterodimers. Isoform 2: Acts as a negative regulator of heterotrimeric G proteins assembly by trapping the preloaded G beta subunits inside the CCT chaperonin.

Type: Primary
Antigen: PHLP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/301
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR