Anti-PDCL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Bioss
BS-5812R-FITC
BOSSBS-5812R-FITCEA 10550 NOK
BOSSBS-5812R-FITC
Anti-PDCL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Isoform 1: Functions as a co-chaperone for CCT in the assembly of heterotrimeric G protein complexes, facilitates the assembly of both Gbeta-Ggamma and RGS-Gbeta5 heterodimers. Isoform 2: Acts as a negative regulator of heterotrimeric G proteins assembly by trapping the preloaded G beta subunits inside the CCT chaperonin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDCL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR