Anti-ADAM11 Rabbit Polyclonal Antibody (Cy7®)
Leverandør: Bioss
BS-5846R-CY7
BOSSBS-5846R-CY7EA 10550 NOK
BOSSBS-5846R-CY7
Anti-ADAM11 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
ADAM11 was first described as MDC (Metalloproteinase-like disintergin-like cysteine-rich protein) from analysis of human brain libraries, in search of brain-specific proteins. Two splice variants with different carboxyterminal ends were described. The message was found only in the brain in this publication. Another group identified ADAM11 in the human brain, where ADAM11 was thought to be involved in cell migration and spatial patterning. ADAM11 was mapped to 17q21.3, a region of interest for breast cancer, and mutations in ADAM11 are associated with some breast cancers. Retinoic acid causedMore Product Information