Anti-ADAMTSL3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-5863R-A555
BOSSBS-5863R-A555EA 10450 NOK
BOSSBS-5863R-A555
Anti-ADAMTSL3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Expressed in epithelial cells of the colon, fallopian tube, skin, breast, prostate, epididymis, liver, pancreatic islets and bile ducts, as well as by vascular endothelial cells, smooth muscle cells, fibroblasts, cortical and ganglionic neurons and cardiac myocytes. Also expressed by malignant epithelial cells in colon cancer, as well as breast, prostate, renal and skin tumors. Expression is significantly reduced in colon cancer compared to normal colon.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADAMTSL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR