Anti-MUC7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-5879R-A750
BOSSBS-5879R-A750EA 10425 NOK
BOSSBS-5879R-A750
Anti-MUC7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
May function in a protective capacity by promoting the clearance of bacteria in the oral cavity and aiding in mastication, speech, and swallowing. Binds P.aeruginosa pili.

Type: Primary
Antigen: MUC7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/377
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR