Anti-CADM4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-5997R-A350
BOSSBS-5997R-A350EA 10450 NOK
BOSSBS-5997R-A350
Anti-CADM4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
Involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CADM4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol CADM4
Antigen-navn Cell Adhesion Molecule 4
Environmentally Preferable
Antigen synonymer Cell adhesion molecule 4|CADM4_HUMAN|IgSF4C|Cadm4|Nectin-like protein 4|TSLC1-like protein 2.|Immunoglobulin superfamily member 4C|NECL-4
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 350
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 199731
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR