Anti-CADM4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverandør: Bioss
BS-5997R-A647
BOSSBS-5997R-A647EA 10450 NOK
BOSSBS-5997R-A647
Anti-CADM4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
Involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CADM4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR