Anti-CADM 4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-5997R-A680
BOSSBS-5997R-A680EA 10450 NOK
BOSSBS-5997R-A680
Anti-CADM 4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity.

Type: Primary
Antigen: CADM 4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 141-240/388
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR