Anti-CADM 4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-5997R-A750
BOSSBS-5997R-A750EA 10450 NOK
BOSSBS-5997R-A750
Anti-CADM 4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity.

Type: Primary
Antigen: CADM 4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 141-240/388
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn CADM 4
Antigen synonymer cadm4|Cell adhesion molecule 4|CADM4_HUMAN|IgSF4C|Nectin-like protein 4|TSLC1-like protein 2|Immunoglobulin superfamily member 4C|NECL-4
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet
Vert Rabbit
Gen-ID 199731
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 141-240/388
Immunogen KLH conjugated synthetic peptide derived from human CADM 4
Skjema Liquid
Molekylvekt 40 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR