Anti-CADM4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Bioss
BS-5997R-HRP
BOSSBS-5997R-HRPEA 10550 NOK
BOSSBS-5997R-HRP
Anti-CADM4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CADM4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR