Anti-HPGD Rabbit Polyclonal Antibody (Cy5.5®)

Leverandør: Bioss
BS-6051R-CY5.5
BOSSBS-6051R-CY5.5EA 10725 NOK
BOSSBS-6051R-CY5.5
Anti-HPGD Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HPGD
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR