Anti-PDGFRL Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-6081R
BOSSBS-6081REA 8500 NOK
BOSSBS-6081R
Anti-PDGFRL Rabbit Polyclonal Antibody
Antistoffer
This protein is expressed in colon, lung and liver. Defects in PDGFRL are associated with colorectal cancer.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PDGFRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR