Anti-PDGFRL Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-6081R-A750
BOSSBS-6081R-A750EA 10450 NOK
BOSSBS-6081R-A750
Anti-PDGFRL Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
This protein is expressed in colon, lung and liver. Defects in PDGFRL are associated with colorectal cancer.

Type: Primary
Antigen: PDGFRL
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 61-150/375
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn PDGFRL
Antigen synonymer PDGFR-like protein|PRLTS|PDGRL|PGFRL_HUMAN|PDGF receptor beta like tumor suppressor
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet
Vert Rabbit
Gen-ID 5157
Isotype IgG
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 61-150/375
Immunogen KLH conjugated synthetic peptide derived from human PDGFRL
Skjema Liquid
Molekylvekt 40 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR