Anti-PDGFRL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Bioss
BS-6081R-FITC
BOSSBS-6081R-FITCEA 10550 NOK
BOSSBS-6081R-FITC
Anti-PDGFRL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
This protein is expressed in colon, lung and liver. Defects in PDGFRL are associated with colorectal cancer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDGFRL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR