Anti-DRES17 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-6299R-A750
BOSSBS-6299R-A750EA 10450 NOK
BOSSBS-6299R-A750
Anti-DRES17 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
PRUNE, the human homologue of the Drosophila gene, is located in 1q21.3, a region highly amplified in human sarcomas, malignant tumours of mesenchymal origin. Human prune (h-prune), a phosphoesterase DHH family appertaining protein, physically interacts with nm23-H1, a metastasis suppressor gene. h-prune is involved in cellular motility and metastasis formation. Metastatic breast cancers were found to overexpress h-prune; h-prune was also found to be highly expressed in colorectal and pancreatic cancers. Hence h-prune is considered useful as a marker for tumor aggressiveness.

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn DRES17
Antigen synonymer hprune|HTCD37|PRUNE_HUMAN|PRUNE like protein|TcD37 homolog|Prune homolog|DRES17|Drosophila-related expressed sequence 17
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet Mouse
Vert Rabbit
Gen-ID 58497
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 201-300/453
Immunogen KLH conjugated synthetic peptide derived from human DRES17
Skjema Liquid
Molekylvekt 50 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR