Anti-PRUNE Rabbit Polyclonal Antibody (Cy5.5®)
Leverandør: Bioss
BS-6299R-CY5.5
BOSSBS-6299R-CY5.5EA 10550 NOK
BOSSBS-6299R-CY5.5
Anti-PRUNE Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
PRUNE, the human homologue of the Drosophila gene, is located in 1q21.3, a region highly amplified in human sarcomas, malignant tumours of mesenchymal origin. Human prune (h-prune), a phosphoesterase DHH family appertaining protein, physically interacts with nm23-H1, a metastasis suppressor gene. h-prune is involved in cellular motility and metastasis formation. Metastatic breast cancers were found to overexpress h-prune; h-prune was also found to be highly expressed in colorectal and pancreatic cancers. Hence h-prune is considered useful as a marker for tumor aggressiveness.
RecommendedMore Product Information