Anti-Adenylosuccinate Lyase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Leverandør: Bioss
BS-6352R-A750
BOSSBS-6352R-A750EA 10450 NOK
BOSSBS-6352R-A750
Anti-Adenylosuccinate Lyase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Adenylsuccinate lyase is involved in both de novo synthesis of purines and formation of adenosine monophosphate from inosine monophosphate. It catalyzes two reactions in AMP biosynthesis: the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and removal of fumarate from adenylosuccinate to give AMP. Adenylosuccinase deficiency results in succinylpurinemic autism, psychomotor retardation, and , in some cases, growth retardation associated with muscle wasting and epilepsy. Two transcript variants encoding differentMore Product Information