Anti-Ag85B Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-6460R
BOSSBS-6460REA 8425 NOK
BOSSBS-6460R
Anti-Ag85B Rabbit Polyclonal Antibody
Antistoffer
Antigen 85B is the most abundant protein expressed by Mycobacterium tuberculosis (about one quarter). It is a myocolyc transferase in the myc pathway and catalyses - like Ag85A and Ag85C - the transfer of the fatty acid mycolate from one trehalose monomycolate to another, resulting in trehalose dimycolate and free trehalose and helping build the cell wall.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Ag85B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestill nå

SPESIFIKASJONER

Antigensymbol Ag85B
Antigen-navn Antigen 85 complex B
Antigen synonymer Antigen 85-B|Mycobacterium|Mycolyl transferase 85B|Ag85B|Antigen 85 complex B|Extracellular alpha-antigen|fbpB|85B|30 kDa extracellular protein|Rv1886c|MT1934|Fibronectin-binding protein B|MTCY180.32.
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Bacteria
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A.
Konsentrasjon 1 ug/ul
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR