Anti-LRRN2 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-6545R
BOSSBS-6545REA 8500 NOK
BOSSBS-6545R
Anti-LRRN2 Rabbit Polyclonal Antibody
Antistoffer
The protein encoded by this gene belongs to the leucine-rich repeat superfamily. This gene was found to be amplified and overexpressed in malignant gliomas. The encoded protein has homology with other proteins that function as cell-adhesion molecules or as signal transduction receptors and is a candidate for the target gene in the 1q32.1 amplicon in malignant gliomas. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: LRRN2
Clonality: Polyclonal
Clone:
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR