Anti-OCC1 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-6556R
BOSSBS-6556REA 8500 NOK
BOSSBS-6556R
Anti-OCC1 Rabbit Polyclonal Antibody
Antistoffer
OCC1 was initially identified as a novel human cDNA overexpressed in a colon carcinoma cell line. Elevated levels of OCC1 mRNA were observed in three of eight colon carcinomas, suggesting that OCC1 may be a hallmark of only a subset of colon carcinomas. It is highly expressed in skeletal muscle, kidney, placenta, and pancreas tissues, with low to no expression in heart, brain, lung and liver tissues. At least two different OCC1 mRNA species are known to exist.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: OCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR