Anti-HSD17B6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-6592R-A680
BOSSBS-6592R-A680EA 10450 NOK
BOSSBS-6592R-A680
Anti-HSD17B6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
NAD-dependent oxidoreductase with broad substrate specificity that shows both oxidative and reductive activity (in vitro). Has 17-beta-hydroxysteroid dehydrogenase activity towards various steroids (in vitro). Converts 5-alpha-androstan-3-alpha,17-beta-diol to androsterone and estradiol to estrone (in vitro). Has 3-alpha-hydroxysteroid dehydrogenase activity towards androsterone (in vitro). Has retinol dehydrogenase activity towards all-trans-retinol (in vitro). Can convert androsterone to epi-androsterone. Androsterone is first oxidised to 5-alpha-androstane-3,17-dione and then reduced toMore Product Information