Anti-HSD17B6 Rabbit Polyclonal Antibody (Cy5.5®)

Leverandør: Bioss
BS-6592R-CY5.5
BOSSBS-6592R-CY5.5EA 10550 NOK
BOSSBS-6592R-CY5.5
Anti-HSD17B6 Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
NAD-dependent oxidoreductase with broad substrate specificity that shows both oxidative and reductive activity (in vitro). Has 17-beta-hydroxysteroid dehydrogenase activity towards various steroids (in vitro). Converts 5-alpha-androstan-3-alpha,17-beta-diol to androsterone and estradiol to estrone (in vitro). Has 3-alpha-hydroxysteroid dehydrogenase activity towards androsterone (in vitro). Has retinol dehydrogenase activity towards all-trans-retinol (in vitro). Can convert androsterone to epi-androsterone. Androsterone is first oxidized to 5-alpha-androstane-3,17-dione and then reduced toMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol HSD17B6
Environmentally Preferable
Antigen-navn Hydroxysteroid (17-beta) Dehydrogenase 6
Antigen synonymer SDR9C6|HSE|RODH
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5.5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 8630
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR