Anti-RD1 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-6877R
BOSSBS-6877REA 8500 NOK
BOSSBS-6877R
Anti-RD1 Rabbit Polyclonal Antibody
Antistoffer
Could play a role in regulating gene activity in the proliferative and/or differentiative pathways induced by NGF. May be an autocrine factor that attenuates or amplifies the initial ligand-induced signal.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: RD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol RD1
Environmentally Preferable
Antigen-navn Retinal Degeneration 1
Antigen synonymer PC4|TIS7
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 3475
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A.
Konsentrasjon 1 ug/ul
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR