Anti-SDCCAG8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-7011R-A555
BOSSBS-7011R-A555EA 10450 NOK
BOSSBS-7011R-A555
Anti-SDCCAG8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
The exact function of SDCCAG8 remains unknown. It is expressed in thymus, prostate, testis, ovary, small intestine, colon, mucosa, colon and renal cancer tumors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDCCAG8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol SDCCAG8
Antigen-navn Serologically Defined Colon Cancer Antigen 8
Environmentally Preferable
Antigen synonymer hCCCAP|NY-CO-8|CCCAP SLSN7|HSPC085|NPHP10|CCCAP|BBS16|SLSN7
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 555
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 10806
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR