Anti-SDCCAG8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-7011R-A680
BOSSBS-7011R-A680EA 10450 NOK
BOSSBS-7011R-A680
Anti-SDCCAG8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
The exact function of SDCCAG8 remains unknown. It is expressed in thymus, prostate, testis, ovary, small intestine, colon, mucosa, colon and renal cancer tumors.

Type: Primary
Antigen: SDCCAG8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 321-420/713
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigen-navn SDCCAG8
Environmentally Preferable
Antigen synonymer NPHP10 gene|SDCG8_HUMAN|NY-CO-8|SLSN7 gene|HSPC085|CCCAP|Centrosomal colon cancer autoantigen protein|Serologically defined colon cancer antigen 8|Antigen NY CO 8|Serologically defined colon cancer antigen 8 homolog
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet
Vert Rabbit
Gen-ID 10806
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 321-420/713
Immunogen KLH conjugated synthetic peptide derived from human SDCCAG8
Skjema Liquid
Molekylvekt 83 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR