Anti-DOCK1 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-7110R
BOSSBS-7110REA 8500 NOK
BOSSBS-7110R
Anti-DOCK1 Rabbit Polyclonal Antibody
Antistoffer
Involved in cytoskeletal rearrangements required for phagocytosis of apoptotic cells and cell motility. Functions as a guanine nucleotide exchange factor (GEF), which activates Rac Rho small GTPases by exchanging bound GDP for free GTP. Its GEF activity may be enhanced by ELMO1.Tissue specificity: Highly expressed in placenta, lung, kidney, pancreas and ovary. Expressed at intermediate level in thymus, testes and colon.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DOCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR