Anti-ATP2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-7539R-A555
BOSSBS-7539R-A555EA 10450 NOK
BOSSBS-7539R-A555
Anti-ATP2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen. Contributes to calcium sequestration involved in muscular excitation/contraction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATP2A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR