Anti-SERCA1 ATPase Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-7539R-A750
BOSSBS-7539R-A750EA 10450 NOK
BOSSBS-7539R-A750
Anti-SERCA1 ATPase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen. Contributes to calcium sequestration involved in muscular excitation/contraction.

Type: Primary
Antigen: SERCA1 ATPase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/1001
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR