Anti-ATP2A1 Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-7539R-CY3
BOSSBS-7539R-CY3EA 10550 NOK
BOSSBS-7539R-CY3
Anti-ATP2A1 Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen. Contributes to calcium sequestration involved in muscular excitation/contraction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATP2A1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol ATP2A1
Environmentally Preferable
Antigen-navn ATPase, Ca++ transporting, cardiac muscle, fast twitch 1
Antigen synonymer SERCA1|ATP2A
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy3®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 487
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR