Anti-CIDEA Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-7649R
BOSSBS-7649REA 8500 NOK
BOSSBS-7649R
Anti-CIDEA Rabbit Polyclonal Antibody
Antistoffer
Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase. DFF45/ICARD has been identified as inhibitor of caspase activated DNase DFF40/CAD. DFF45 related proteins CIDE A and CIDE B (for cell death inducing DFF like effector A and B) were recently identified. CIDE contains a new type of domain termed CIDE N, which has high homologyMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol CIDEA
Antigen-navn Cell death-inducing DFFA-like effector a
Environmentally Preferable
Antigen synonymer Cell death activator CIDE-A|CIDE-A|Cell death-inducing DFFA-like effector A
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 1149
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A.
Konsentrasjon 1 ug/ul
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR