Anti-CIDEA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverandør: Bioss
BS-7649R-A488
BOSSBS-7649R-A488EA 10450 NOK
BOSSBS-7649R-A488
Anti-CIDEA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase. DFF45/ICARD has been identified as inhibitor of caspase activated DNase DFF40/CAD. DFF45 related proteins CIDE A and CIDE B (for cell death inducing DFF like effector A and B) were recently identified. CIDE contains a new type of domain termed CIDE N, which has high homologyMore Product Information