Anti-DNase 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-7651R-A680
BOSSBS-7651R-A680EA 10450 NOK
BOSSBS-7651R-A680
Anti-DNase 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Deoxyribonuclease I gene is approximately 3.2 kb long with 9 exons separated by 8 introns. In the form of a bovine pancreatic enzyme preparation, it occupies an important place in the history of protein chemistry and enzymology: it was the first enzyme to be recognised as specific for DNA; it was the first DNase to be crystallised; and it was the first DNase for which a specific protein inhibitor was characterised. DNase I is a Ca2+ and Mg2+ dependant endonuclease. DNase I is synthesised in the pancreas and stored in zymogen granules. It has been used to reduce the viscosity of cysticMore Product Information